- Epsilon-Polylysine Fermentation and its Recovery Using Carboxymethyl Cellulose (CMC)-Conjugated Magnetite
Epsilon-Polylysine Fermentation and its Recovery Using Carboxymethyl Cellulose (CMC)-Conjugated Magnetite
Separation Science and Technology (2013)
Wibowo N, et al.