Merck
  • Properties of collagen and hyaluronic acid composite materials and their modification by chemical crosslinking
Properties of collagen and hyaluronic acid composite materials and their modification by chemical crosslinking
Journal of Biomedical Materials Research (1996)
Rehakova M, et al.