Merck
  • The crucial role of protein phosphorylation in cell signaling and its use as targeted therapy
The crucial role of protein phosphorylation in cell signaling and its use as targeted therapy
International Journal of Molecular Medicine (2017)
Ardito F, et al.