- An integrated assessment of histopathological changes of the enteric neuromuscular compartment in experimental colitis
An integrated assessment of histopathological changes of the enteric neuromuscular compartment in experimental colitis
Journal of Cellular and Molecular Medicine (2015)
Ippolito C, et al.