- Transplantation of bone marrow-derived MSCs improves cisplatinum-induced renal injury through paracrine mechanisms
Transplantation of bone marrow-derived MSCs improves cisplatinum-induced renal injury through paracrine mechanisms
Experimental and Molecular Pathology (2013)
Cheng K, et al.