Merck
  • High performance liquid chromatography of acarbose and its metabolite on porous graphitic carbon column.
High performance liquid chromatography of acarbose and its metabolite on porous graphitic carbon column.
Journal of Separation Science (2002)
Daali Y, et al.