Merck
  • N-containing boronic esters as self-complementary building blocks for the assembly of 2D and 3D molecular networks.
N-containing boronic esters as self-complementary building blocks for the assembly of 2D and 3D molecular networks.
Chemical communications (Cambridge, England) (2012-03-23)
Jorge Cruz-Huerta, Domingo Salazar-Mendoza, Javier Hernández-Paredes, Irán F Hernández Ahuactzi, Herbert Höpfl
ABSTRACT
The combination of two heteroaromatic boronic acids with pentaerythritol gave self-complementary tectons which were suitable for the generation of 2D and 3D molecular networks.
Sigma-Aldrich
4-Pyridinylboronic acid, 90%
Pricing and availability is not currently available.