Merck
  • Stomatin-like protein 2 inhibits cisplatin-induced apoptosis through MEK/ERK signaling and the mitochondrial apoptosis pathway in cervical cancer cells.
Stomatin-like protein 2 inhibits cisplatin-induced apoptosis through MEK/ERK signaling and the mitochondrial apoptosis pathway in cervical cancer cells.
Cancer science (2018-03-09)
Guolin Hu, Jialu Zhang, Feifei Xu, Huan Deng, Weiwei Zhang, Shijun Kang, Weijiang Liang
ABSTRACT
Stomatin-like protein 2 (STOML2 or SLP-2) is an oncogenic anti-apoptotic protein that is upregulated in several types of cancer, including cervical cancer. However, the mechanisms responsible for the SLP-2 anti-apoptotic function remain poorly understood. Here, we show that siRNA-mediated SLP-2 suppression decreases growth of human cervical cancer HELA and SIHA cells, and increases cisplatin-induced apoptosis through activation of MEK/ERK signaling and suppression of the mitochondrial pathway. The inhibition of the mitochondrial pathway is mediated by increasing the mitochondrial Ca2+ concentration and mitochondrial membrane potential, thereby downregulating p-MEK and p-ERK levels, upregulating the Bax/Bcl-2 ratio, increasing cytochrome C release from mitochondria into the cytosol, and upregulating levels of cleaved-caspase 9, cleaved-caspase 3, and cleaved poly ADP-ribose polymerase (PARP). Overexpression of SLP-2 using adenovirus-STOML2 has the opposite effect: it upregulates p-MEK and p-ERK and downregulates the Bax/Bcl-2 ratio and levels of cleaved-caspase 9 to caspase 9, cleaved-caspase 3 to caspase 3, and cleaved-PARP to PARP in cisplatin-treated cells. These data show that SLP-2 inhibits cisplatin-induced apoptosis by activating the MEK/ERK signaling and inhibiting the mitochondrial apoptosis pathway in cervical cancer cells.