Merck
  • ZBTB10 binds the telomeric variant repeat TTGGGG and interacts with TRF2.
ZBTB10 binds the telomeric variant repeat TTGGGG and interacts with TRF2.
Nucleic acids research (2019-01-11)
Alina Bluhm, Nikenza Viceconte, Fudong Li, Grishma Rane, Sandra Ritz, Suman Wang, Michal Levin, Yunyu Shi, Dennis Kappei, Falk Butter
ABSTRACT
Telomeres are nucleoprotein structures at the ends of linear chromosomes and present an essential feature for genome integrity. Vertebrate telomeres usually consist of hexameric TTAGGG repeats, however, in cells that use the alternative lengthening of telomeres (ALT) mechanism, variant repeat sequences are interspersed throughout telomeres. Previously, it was shown that NR2C/F transcription factors bind to TCAGGG variant repeats and contribute to telomere maintenance in ALT cells. While specific binders to other variant repeat sequences have been lacking to date, we here identify ZBTB10 as the first TTGGGG-binding protein and demonstrate direct binding via the two zinc fingers with affinity in the nanomolar range. Concomitantly, ZBTB10 co-localizes with a subset of telomeres in ALT-positive U2OS cells and interacts with TRF2/RAP1 via the N-terminal region of TRF2. Our data establishes ZBTB10 as a novel variant repeat binding protein at ALT telomeres.
Sigma-Aldrich
RO-3306, ≥98% (HPLC)