Merck
  • Square-wave voltammetric (SWV) determination of Captopril in reconstituted serum and pharmaceutical formulations
Square-wave voltammetric (SWV) determination of Captopril in reconstituted serum and pharmaceutical formulations
Talanta (2005)
Parham H and Zargar B