- Determination of captopril and its degradation products by capillary electrophoresis
Determination of captopril and its degradation products by capillary electrophoresis
Journal of Pharmaceutical and Biomedical Analysis (1999)
Hillaert S and Van den Bossche W