Merck
  • Determination of Ofloxacin in Pharmaceutical Forms by High-Performance Liquid Chromatography and Derivative Uvspectrophotometry
Determination of Ofloxacin in Pharmaceutical Forms by High-Performance Liquid Chromatography and Derivative Uvspectrophotometry
Analytical Letters (1993)
Carlucci G, et al.