Merck
  • Defining standard enzymatic dissociation methods for individual brains and spinal cords in EAE
Defining standard enzymatic dissociation methods for individual brains and spinal cords in EAE
Neurology? neuroimmunology & neuroinflammation (2018)
Hussain RZ, et al.