Merck
  • Pathogen reduction technology treatment of platelets, plasma and whole blood using riboflavin and UV light
Pathogen reduction technology treatment of platelets, plasma and whole blood using riboflavin and UV light
Transfusion Medicine and Hemotherapy (2011)