- Blocking the mineralocorticoid receptor improves effectiveness of steroid treatment for low back pain in rats
Blocking the mineralocorticoid receptor improves effectiveness of steroid treatment for low back pain in rats
Anesthesiology (2014)
Ye L, et al.