Merck
  • Triphenylphosphine oxide-catalyzed stereoselective poly-and dibromination of unsaturated compounds
Triphenylphosphine oxide-catalyzed stereoselective poly-and dibromination of unsaturated compounds
Chemical Communications (Cambridge, England) (2014)
Yu T, et al.