- An amino-modified Zr-terephthalate metal?organic framework as an acid?base catalyst for cross-aldol condensation
An amino-modified Zr-terephthalate metal?organic framework as an acid?base catalyst for cross-aldol condensation
Chemical Communications (Cambridge, England) (2011)
Vermoortele F, et al.