Merck
  • Synthesis and characterization of cycloaliphatic hydrophilic polyurethanes, modified with L-ascorbic acid, as materials for soft tissue regeneration
Synthesis and characterization of cycloaliphatic hydrophilic polyurethanes, modified with L-ascorbic acid, as materials for soft tissue regeneration
Materials Science & Engineering. C, Materials For Biological Applications (2017)
Kucinska-Lipka J, et al.