Merck
  • Methyl-Specific Isotope Labeling Strategies for NMR Studies of Membrane Proteins.
Methyl-Specific Isotope Labeling Strategies for NMR Studies of Membrane Proteins.
Methods in molecular biology (Clifton, N.J.) (2017-07-30)
Vilius Kurauskas, Paul Schanda, Remy Sounier
ABSTRACT
Methyl groups are very useful probes of structure, dynamics, and interactions in protein NMR spectroscopy. In particular, methyl-directed experiments provide high sensitivity even in very large proteins, such as membrane proteins in a membrane-mimicking environment. In this chapter, we discuss the approach for labeling methyl groups in E. coli-based protein expression, as exemplified with the mitochondrial carrier GGC.
Sigma-Aldrich
QLAM-AβIδ1LVproR-U-13C Methyl Labeling Kit