Merck
  • Inhibition of substance-P-induced histamine release from rat peritoneal mast cells by 8-methoxypsoralen plus long-wave ultraviolet light irradiation: decreased intracellular calcium as a possible mechanism.
Inhibition of substance-P-induced histamine release from rat peritoneal mast cells by 8-methoxypsoralen plus long-wave ultraviolet light irradiation: decreased intracellular calcium as a possible mechanism.
International archives of allergy and immunology (1998-01-16)
H Amano, M Kurosawa, Y Miyachi
ABSTRACT
Rat peritoneal mast cells purified on a Percoll gradient were activated by substance P (SP) and the effect of 8-methoxypsoralen (8-MOP) plus long-wave ultraviolet light (UVA) irradiation (8-MOP/UVA) on SP-induced histamine release from the cells was investigated. At a concentration of 10(-5) M SP caused a significant histamine release. 8-MOP or UVA irradiation alone at the doses used in the present study neither induced a histamine release nor had any significant effects on SP-induced histamine release from mast cells. On the other hand, 8-MOP/UVA inhibited SP-induced histamine release; this inhibition was dependent on the UVA doses (0.5-3.0 J/cm2) and was accompanied by a reduction in the rise in intracellular calcium concentrations ([Ca2+]i). These data suggest that 8-MOP/UVA inhibits histamine release from SP-activated rat peritoneal mast cells by suppressing the rise in [Ca2+]i.
Product Description
Supelco
8-Methoxypsoralen, analytical standard