Merck
  • Molecularly Imprinted Sol-Gel for TNT Detection with Optical Micro-Ring Resonator Sensor Chips.
Molecularly Imprinted Sol-Gel for TNT Detection with Optical Micro-Ring Resonator Sensor Chips.
Sensors (Basel, Switzerland) (2019-09-13)
Ludmila Eisner, Isabel Wilhelm, Günter Flachenecker, Jürgen Hürttlen, Wolfgang Schade
ABSTRACT
A sensor for trinitrotoluene (TNT) detection was developed by using a combination of optical micro-ring technology and a receptor coating based on molecularly imprinted sol-gel layers. Two techniques for deposition of receptor layers were compared: Airbrush technology and electrospray ionization. A concentration of less than 5 ppb for TNT in the gas-phase, using electrospray deposition of the receptor layer, was detected. The cross-sensitivities to organic substances and further nitro-based explosives were compared. As a result, the sensitivity to TNT is about one order of magnitude higher in comparison to the explosives 2,4-dinitrotoluene (DNT) or 1,3-dinitrobenzene (DNB) and about four orders of magnitude higher than the organic substances phenol, ethanol, and acetone. The signal response of the sensor is fast, and the compact sensor design enables the deposition of different receptor layers on multiple optical micro-rings on one chip, which allows a more precise analysis and reduction of side effects and false alarms.
Sigma-Aldrich
3,5-Dinitrobenzyl alcohol, 98%
Pricing and availability is not currently available.