Merck
  • A novel specific peroxisome proliferator-activated receptor γ (PPARγ) modulator YR4-42 ameliorates hyperglycaemia and dyslipidaemia and hepatic steatosis in diet-induced obese mice.
A novel specific peroxisome proliferator-activated receptor γ (PPARγ) modulator YR4-42 ameliorates hyperglycaemia and dyslipidaemia and hepatic steatosis in diet-induced obese mice.
Diabetes, obesity & metabolism (2019-08-01)
Yi Huan, Xuan Pan, Jun Peng, Chunming Jia, Sujuan Sun, Guoliang Bai, Xing Wang, Tian Zhou, Rongcui Li, Shuainan Liu, Caina Li, Quan Liu, Zhanzhu Liu, Zhufang Shen
ABSTRACT
To evaluate a novel tetrahydroisoquinoline derivative YR4-42 as a selective peroxisome proliferator-activated receptor γ (PPARγ) modulator (SPPARM) and explore its anti-diabetic effects in vitro and in vivo. Using two standard full PPARγ agonists rosiglitazone and pioglitazone as controls, the PPARγ binding affinity and transactivation action of YR4-42 were evaluated using biochemical and cell-based reporter gene assays. The capacity of YR4-42 to recruit coactivators of PPARγ was also assessed. The effects of YR4-42 on adipogenesis and glucose consumption and PPARγ Ser273 phosphorylation were investigated in 3T3-L1 adipocytes. The effects of YR4-42 and pioglitazone, serving as positive control, on glucose and lipids metabolism were investigated in high-fat diet-induced obese (DIO) C57BL/6J mice. The expression of PPARγ target genes involved in glucose and lipid metabolism was also assessed in vitro and in vivo. In vitro biochemical and cell-based functional assays showed that YR4-42 has much weaker binding affinity, transactivation, and recruitment to PPARγ of the coactivators thyroid hormone receptor-associated protein complex 220 kDa component (TRAP220) and PPARγ coactivator 1-α (PGC1α) compared to full agonists. In 3 T3-L1 adipocytes, YR4-42 significantly improved glucose consumption without a lipogenesis effect, while blocking tumour necrosis factor α-mediated phosphorylation of PPARγ at Ser273, thereby upregulating the expression of the PPARγ Ser273 phosphorylation-dependent genes. Furthermore, in DIO mice, oral administration of YR4-42 ameliorated the hyperglycaemia, with a similar insulin sensitization effect to that of pioglitazone. Importantly, YR4-42 also improved hyperlipidaemia-associated hepatic steatosis without weight gain, which avoids a major side effect of pioglitazone. Thus, YR4-42 appeared to selectively modulate PPARγ responses. This finding was supported by the gene expression analysis, which showed that YR4-42 selectively targets PPARγ-regulated genes mapped to glucose and lipid metabolism in DIO mice. We conclude that YR4-42 is a novel anti-diabetic drug candidate with significant advantages compared to standard PPARγ agonists. YR4-42 should be further investigated in preclinical and clinical studies.
Sigma-Aldrich
GW7647, ≥98% (HPLC)