- Synthesis and characterization of triblock copolymers of methoxy poly (ethylene glycol) and poly (propylene fumarate)
Synthesis and characterization of triblock copolymers of methoxy poly (ethylene glycol) and poly (propylene fumarate)
Biomacromolecules (2002)
Behravesh E, et al.