Merck
  • Physicochemical properties and storage stability of spray-dried soya bean sprout extract
Physicochemical properties and storage stability of spray-dried soya bean sprout extract
International Journal of Food Science and Technology (2016)
Lee D, et al.