Merck
  • Strain-dependent genomic factors affect allergen-induced airway hyperresponsiveness in mice.
Strain-dependent genomic factors affect allergen-induced airway hyperresponsiveness in mice.
American journal of respiratory cell and molecular biology (2011-03-08)
Samir N P Kelada, Mark S Wilson, Urraca Tavarez, Kari Kubalanza, Bhavesh Borate, Greg S Whitehead, Shuichiro Maruoka, Michelle G Roy, Michelle Olive, Danielle E Carpenter, David M Brass, Thomas A Wynn, Donald N Cook, Christopher M Evans, David A Schwartz, Francis S Collins
ABSTRACT
Asthma is etiologically and clinically heterogeneous, making the genomic basis of asthma difficult to identify. We exploited the strain-dependence of a murine model of allergic airway disease to identify different genomic responses in the lung. BALB/cJ and C57BL/6J mice were sensitized with the immunodominant allergen from the Dermatophagoides pteronyssinus species of house dust mite (Der p 1), without exogenous adjuvant, and the mice then underwent a single challenge with Der p 1. Allergic inflammation, serum antibody titers, mucous metaplasia, and airway hyperresponsiveness were evaluated 72 hours after airway challenge. Whole-lung gene expression analyses were conducted to identify genomic responses to allergen challenge. Der p 1-challenged BALB/cJ mice produced all the key features of allergic airway disease. In comparison, C57BL/6J mice produced exaggerated Th2-biased responses and inflammation, but exhibited an unexpected decrease in airway hyperresponsiveness compared with control mice. Lung gene expression analysis revealed genes that were shared by both strains and a set of down-regulated genes unique to C57BL/6J mice, including several G-protein-coupled receptors involved in airway smooth muscle contraction, most notably the M2 muscarinic receptor, which we show is expressed in airway smooth muscle and was decreased at the protein level after challenge with Der p 1. Murine strain-dependent genomic responses in the lung offer insights into the different biological pathways that develop after allergen challenge. This study of two different murine strains demonstrates that inflammation and airway hyperresponsiveness can be decoupled, and suggests that the down-modulation of expression of G-protein-coupled receptors involved in regulating airway smooth muscle contraction may contribute to this dissociation.
Sigma-Aldrich
Anti-Muscarinic Acetylcholine Receptor m2 Antibody, clone M2-2-B3, clone M2-2-B3, Chemicon®, from rat