Merck
  • NKG2D is critical for NK cell activation in host defense against Pseudomonas aeruginosa respiratory infection
NKG2D is critical for NK cell activation in host defense against Pseudomonas aeruginosa respiratory infection
Journal of immunology (Baltimore, Md. : 1950) (2008)
Wesselkamper S, et al.