Merck
  • Peloruside- and laulimalide-resistant human ovarian carcinoma cells have βI-tubulin mutations and altered expression of βII- and βIII-tubulin isotypes.
Peloruside- and laulimalide-resistant human ovarian carcinoma cells have βI-tubulin mutations and altered expression of βII- and βIII-tubulin isotypes.
Molecular cancer therapeutics (2011-06-10)
Arun Kanakkanthara, Anja Wilmes, Aurora O'Brate, Daniel Escuin, Ariane Chan, Ada Gjyrezi, Janet Crawford, Pisana Rawson, Bronwyn Kivell, Peter T Northcote, Ernest Hamel, Paraskevi Giannakakou, John H Miller
ABSTRACT
Peloruside A and laulimalide are potent microtubule-stabilizing natural products with a mechanism of action similar to that of paclitaxel. However, the binding site of peloruside A and laulimalide on tubulin remains poorly understood. Drug resistance in anticancer treatment is a serious problem. We developed peloruside A- and laulimalide-resistant cell lines by selecting 1A9 human ovarian carcinoma cells that were able to grow in the presence of one of these agents. The 1A9-laulimalide resistant cells (L4) were 39-fold resistant to the selecting agent and 39-fold cross-resistant to peloruside A, whereas the 1A9-peloruside A resistant cells (R1) were 6-fold resistant to the selecting agent while they remained sensitive to laulimalide. Neither cell line showed resistance to paclitaxel or other drugs that bind to the taxoid site on β-tubulin nor was there resistance to microtubule-destabilizing drugs. The resistant cells exhibited impaired peloruside A/laulimalide-induced tubulin polymerization and impaired mitotic arrest. Tubulin mutations were found in the βI-tubulin isotype, R306H or R306C for L4 and A296T for R1 cells. This is the first cell-based evidence to support a β-tubulin-binding site for peloruside A and laulimalide. To determine whether the different resistance phenotypes of the cells were attributable to any other tubulin alterations, the β-tubulin isotype composition of the cells was examined. Increased expression of βII- and βIII-tubulin was observed in L4 cells only. These results provide insight into how alterations in tubulin lead to unique resistance profiles for two drugs, peloruside A and laulimalide, that have a similar mode of action.