Merck
  • Histamine inhibits lipopolysaccharide-induced tumor necrosis factor-alpha production in an intercellular adhesion molecule-1- and B7.1-dependent manner.
Histamine inhibits lipopolysaccharide-induced tumor necrosis factor-alpha production in an intercellular adhesion molecule-1- and B7.1-dependent manner.
The Journal of pharmacology and experimental therapeutics (2003-01-23)
Toshihiko Morichika, Hideo Kohka Takahashi, Hiromi Iwagaki, Tadashi Yoshino, Ryuji Tamura, Minori Yokoyama, Shuji Mori, Tadaatsu Akagi, Masahiro Nishibori, Noriaki Tanaka
ABSTRACT
Lipopolysaccharide (LPS) is recognized as a key molecule in the pathogenesis of Gram negative sepsis and septic shock. In the present study, we demonstrate that LPS (1-1000 pg/ml) concentration dependently up-regulated the expression of intercellular adhesion molecule (ICAM)-1, B7.1, and B7.2 on human monocytes using fluorescence-activated cell sorting analysis, and that tumor necrosis factor (TNF)-alpha production induced by LPS in peripheral blood mononuclear cells (PBMCs) was inhibited by the addition of antibodies against these adhesion molecules, suggesting the dependence of TNF-alpha production on cell-cell interaction through these adhesion molecules. Moreover, we found that histamine (10(-7)-10(-4) M) concentration dependently inhibited the expression of ICAM-1 and B7.1, but not B7.2 on monocytes induced by LPS. Histamine also inhibited the responses of TNF-alpha production induced by LPS. The modulatory effects of histamine on ICAM-1 and B7.1 expression and TNF-alpha production were all concentration dependently antagonized by famotidine but not by d-chlorpheniramine and thioperamide, and were mimicked by selective H2-receptor agonists but not by H1-, H3-, and H4-receptor agonists, indicating the involvement of H2-receptors in the histamine action. Dibutyryl cAMP down-regulated ICAM-1 and B7.1 expression on monocytes stimulated by LPS, suggesting the mediation by the cyclic adenosine monophosphate-protein kinase A pathway of H2-receptor activation. These results as a whole indicated that histamine via H2-receptor inhibited the LPS-induced TNF-alpha production through the regulation of ICAM-1 and B7.1 expression, leading to the reduction of innate immune response stimulated by LPS.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O26:B6, ≥10,000 EU/mg, purified by phenol extraction