Merck
  • Type 2 diabetes-associated single nucleotide polymorphism in Sorcs1 gene results in alternative processing of the Sorcs1 protein in INS1 β-cells.
Type 2 diabetes-associated single nucleotide polymorphism in Sorcs1 gene results in alternative processing of the Sorcs1 protein in INS1 β-cells.
Scientific reports (2019-12-21)
Belinda Yau, Zachary Blood, Yousun An, Zhiduan Su, Melkam A Kebede
ABSTRACT
A threonine-to-Isoleucine (Thr52Ile) mutation within the pro-domain of the Sorcs1 gene was positionally cloned as the gene underlying a quantitative trait locus that affects fasting insulin levels in mice. In humans, genome-wide association studies and linkage studies have shown that SORCS1 is associated with diabetes and all of diabetes complications. We have recently shown that deletion of Sorcs1 in mice made obese with the leptinob mutation results in diabetes and an insulin granule stability defect. This present study investigates the functional consequence of the Sorcs1 Thr52Ile mutation in the rat INS1 β-cell line expressing either the wildtype or mutant Sorcs1 allele. We find that Sorcs1 Thr52Ile mutation is associated with increased basal insulin secretion, reduced glucose-stimulated insulin secretion and decreased insulin content in INS1 cells. Moreover, expression of Thr52Ile causes differential processing of the Sorcs1 protein resulting in the formation of an additional 90 kDa mutant form of the protein. The mutant form of the protein is localised to the ER, retains its pro-domain, and concurrently reduces expression of the functional mature 130 kDa Sorcs1 protein. These findings provide a mechanistic clue to why this specific allelic variation in Sorcs1 was associated with reduced insulin levels and type 2 diabetes.
Sigma-Aldrich
Anti-TGN38 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Brilliant Blue G, pure
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
Anti-Myc Tag Antibody, clone 4A6, agarose conjugate, clone 4A6, Upstate®, from mouse
Millipore
RIPA Lysis Buffer, 10X, 100 mL RIPA Lysis Buffer, 10X for Immunoprecipitation & Western Blotting.