Merck
  • Methionine deficiency does not increase polyamine turnover through depletion of hepatic S-adenosylmethionine in juvenile Atlantic salmon.
Methionine deficiency does not increase polyamine turnover through depletion of hepatic S-adenosylmethionine in juvenile Atlantic salmon.
The British journal of nutrition (2014-09-10)
Marit Espe, Synne Marte Andersen, Elisabeth Holen, Ivar Rønnestad, Eva Veiseth-Kent, Jens-Erik Zerrahn, Anders Aksnes
ABSTRACT
During the last few decades, plant protein ingredients such as soya proteins have replaced fishmeal in the diets of aquacultured species. This may affect the requirement and metabolism of methionine as soya contains less methionine compared with fishmeal. To assess whether methionine limitation affects decarboxylated S-adenosylmethionine availability and polyamine status, in the present study, juvenile Atlantic salmon were fed a methionine-deficient plant protein-based diet or the same diet supplemented with dl-methionine for 8 weeks. The test diets were compared with a fishmeal-based control diet to assess their effects on the growth performance of fish. Methionine limitation reduced growth and protein accretion, but when fish were fed the dl-methionine-supplemented diet their growth and protein accretion equalled those of fish fed the fishmeal-based control diet. Methionine limitation reduced free methionine concentrations in the plasma and muscle, while those in the liver were not affected. S-adenosylmethionine (SAM) concentrations were higher in the liver of fish fed the methionine-deficient diet, while S-adenosylhomocysteine concentrations were not affected. Putrescine concentrations were higher and spermine concentrations were lower in the liver of fish fed the methionine-deficient diet, while the gene expression of SAM decarboxylase (SAMdc) and the rate-limiting enzyme of polyamine synthesis ornithine decarboxylase (ODC) was not affected. Polyamine turnover, as assessed by spermine/spermidine acetyltransferase (SSAT) abundance, activity and gene expression, was not affected by treatment. However, the gene expression of the cytokine TNF-α increased in fish fed the methionine-deficient diet, indicative of stressful conditions in the liver. Even though taurine concentrations in the liver were not affected by treatment, methionine and taurine concentrations in muscle decreased due to methionine deficiency. Concomitantly, liver phospholipid and cholesterol concentrations were reduced, while NEFA concentrations were elevated. In conclusion, methionine deficiency did not increase polyamine turnover through depletion of hepatic SAM, as assessed by SSAT activity and abundance.
Sigma-Aldrich
Anti-PEMT antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.