Merck
  • Co-assembly of two GluR6 kainate receptor splice variants within a functional protein complex.
Co-assembly of two GluR6 kainate receptor splice variants within a functional protein complex.
Neuron (2005-08-17)
Françoise Coussen, David Perrais, Frédéric Jaskolski, Shankar Sachidhanandam, Elisabeth Normand, Joel Bockaert, Philippe Marin, Christophe Mulle
ABSTRACT
Kainate receptors (KAR) are composed of several distinct subunits and splice variants, but the functional relevance of this diversity remains largely unclear. Here we show that two splice variants of the GluR6 subunit, GluR6a and GluR6b, which differ in their C-terminal domains, do not show distinct functional properties, but coassemble as heteromers in vitro and in vivo. Using a proteomic approach combining affinity purification and MALDI-TOF mass spectrometry, we found that GluR6a and GluR6b interact with two distinct subsets of cytosolic proteins mainly involved in Ca(2+) regulation of channel function and intracellular trafficking. Guided by these results, we provide evidence that the regulation of native KAR function by NMDA receptors depends on the heteromerization of GluR6a and GluR6b and interaction of calcineurin with GluR6b. Thus, GluR6a and GluR6b bring in close proximity two separate subsets of interacting proteins that contribute to the fine regulation of KAR trafficking and function.
Sigma-Aldrich
Anti-Myc Tag Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-Calmodulin Antibody, Upstate®, from mouse
Sigma-Aldrich
Anti-KA2/GRIK5 (Kainate Receptor) Antibody, Upstate®, from rabbit