Merck
TIS11D is a candidate pro-apoptotic p53 target gene.
Cell cycle (Georgetown, Tex.) (2006-12-19)
Roger S Jackson, Yong-Jig Cho, Peng Liang
ABSTRACT
A number of target genes for the tumor suppressor, p53, have been identified, however, the mechanisms that contribute to p53-dependent apoptosis remain to be fully elucidated. In a comprehensive screen for p53 target genes by differential display, we have identified TIS11D as a p53-inducible gene. Induction of TIS11D mRNA was confirmed by Northern Blot in response to p53 expression. Inducible expression of TIS11D resulted in inhibition of cell proliferation and apoptosis. These data suggest TIS11D as a candidate p53 target gene that may be part of the network of genes responsible for p53-dependent apoptosis.
Sigma-Aldrich
Anti-p53 (Ab-2) (Pantropic) Mouse mAb (PAb1801), liquid, clone PAb1801, Calbiochem®
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.