Merck
  • Zinc (II) coordination architectures with two bulky anthracene-based carboxylic ligands: crystal structures and luminescent properties
Zinc (II) coordination architectures with two bulky anthracene-based carboxylic ligands: crystal structures and luminescent properties
CrystEngComm (2008)
Wang J, et al.