Merck
  • Ultramicroporous Metal- Organic Framework Based on 9, 10-Anthracenedicarboxylate for Selective Gas Adsorption
Ultramicroporous Metal- Organic Framework Based on 9, 10-Anthracenedicarboxylate for Selective Gas Adsorption
Inorganic Chemistry (2007)
Ma S, et al.