- Nanostructured Pt decorated graphene and multi walled carbon nanotube based room temperature hydrogen gas sensor
Nanostructured Pt decorated graphene and multi walled carbon nanotube based room temperature hydrogen gas sensor
Nanoscale (2009)
Kaniyoor A, et al.