Merck
  • Retrograde synaptic signaling mediated by K+ efflux through postsynaptic NMDA receptors.
Retrograde synaptic signaling mediated by K+ efflux through postsynaptic NMDA receptors.
Cell reports (2013-11-26)
Pei-Yu Shih, Leonid P Savtchenko, Naomi Kamasawa, Yulia Dembitskaya, Thomas J McHugh, Dmitri A Rusakov, Ryuichi Shigemoto, Alexey Semyanov
ABSTRACT
Synaptic NMDA receptors (NMDARs) carry inward Ca(2+) current responsible for postsynaptic signaling and plasticity in dendritic spines. Whether the concurrent K(+) efflux through the same receptors into the synaptic cleft has a physiological role is not known. Here, we report that NMDAR-dependent K(+) efflux can provide a retrograde signal in the synapse. In hippocampal CA3-CA1 synapses, the bulk of astrocytic K(+) current triggered by synaptic activity reflected K(+) efflux through local postsynaptic NMDARs. The local extracellular K(+) rise produced by activation of postsynaptic NMDARs boosted action potential-evoked presynaptic Ca(2+) transients and neurotransmitter release from Schaffer collaterals. Our findings indicate that postsynaptic NMDAR-mediated K(+) efflux contributes to use-dependent synaptic facilitation, thus revealing a fundamental form of retrograde synaptic signaling.
Sigma-Aldrich
Anti-NMDAR1 Antibody, clone 54.1, clone 54.1, Chemicon®, from mouse