Merck
  • The FcepsilonRIbeta immunoreceptor tyrosine-based activation motif exerts inhibitory control on MAPK and IkappaB kinase phosphorylation and mast cell cytokine production.
The FcepsilonRIbeta immunoreceptor tyrosine-based activation motif exerts inhibitory control on MAPK and IkappaB kinase phosphorylation and mast cell cytokine production.
The Journal of biological chemistry (2004-09-10)
Yasuko Furumoto, Satoshi Nunomura, Tomoyoshi Terada, Juan Rivera, Chisei Ra
ABSTRACT
The high affinity IgE Fc receptor (FcepsilonRI) beta chain functions as a signal amplifier and has been linked to atopy, asthma, and allergy. Herein, we report on a previously unrecognized negative regulatory role for the nonconventional beta chain immunoreceptor tyrosine-based activation motif that contains three tyrosine residues (YX5YX3Y). Degranulation and leukotriene production was found to be impaired in cells expressing the mutated FcepsilonRIbeta immunoreceptor tyrosine-based activation motifs FYY, YYF, FYF, and FFF. In contrast, cytokine synthesis and secretion were enhanced in the YFY and FFF mutants. FcepsilonRI phosphorylation and Lyn kinase co-immunoprecipitation was intact in the YFY mutant but was lost in the FYF and FFF mutants. The phosphorylation of Syk, LAT, phospholipase gamma1/2, and Srchomology 2 domain-containing protein phosphatase 2 was intact, whereas the phosphorylation of SHIP-1 was significantly reduced in the YFY mutant cells. The FYF and FFF mutants were defective in phosphorylating all of these molecules. In contrast, the phosphorylation of ERK, p38 MAPK, IkappaB kinase beta (IKKbeta), and nuclear NFkappaB activity was enhanced in the YFY and FFF mutants. These findings show that the FcepsilonRIbeta functions to both selectively amplify (degranulation and leukotriene secretion) and dampen (lymphokine) mast cell effector responses.
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, Biotin Conjugate, clone 4G10®, Upstate®, from mouse
Pricing and availability is not currently available.