Merck
  • A new ephrin-A1 isoform (ephrin-A1b) with altered receptor binding properties abrogates the cleavage of ephrin-A1a.
A new ephrin-A1 isoform (ephrin-A1b) with altered receptor binding properties abrogates the cleavage of ephrin-A1a.
The Biochemical journal (2003-12-25)
Eivind F Finne, Else Munthe, Hans-Christian Aasheim
ABSTRACT
Ephrins are ligands for the Eph receptor tyrosine kinases, which play important roles in patterning nervous and vascular systems. Ephrin-A1 is a glycosylphosphatidylinositol-anchored ligand that binds to the EphA receptor tyrosine kinases. In the present study, we have identified a new ephrin-A1 isoform, denoted ephrin-A1b (ephrin-A1 isoform b). Compared with the originally described ephrin-A1 sequence, ephrin-A1a [Holzman, Marks and Dixit (1990) Mol. Cell. Biol. 10, 5830-5838], ephrin-A1b lacks a segment of 22 amino acids (residues 131-152). At the transcript level, exon 3 is spliced out in the transcript encoding ephrin-A1b. Transfection of HEK-293T cells (human embryonic kidney 293 cells) with an ephrin-A1b-expressing plasmid resulted in a significant expression of the protein on the cell surface. However, soluble EphA2 receptor (EphA2-Fc) bound weakly to ephrin-A1b-expressing transfectants, but bound strongly to ephrin-A1a-expressing transfectants. Ephrins have been shown to undergo regulated cleavage after interaction with their receptors. This process is inhibited by co-expression of ephrin-A1a and ephrin-A1b, indicating that ephrin-A1b influences the cleavage process. Taken together, these findings indicate that this newly described isoform may regulate the function of its ephrin-A1a counterpart.
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, Biotin Conjugate, clone 4G10®, Upstate®, from mouse
Pricing and availability is not currently available.