Merck
  • 3,4-Dimethoxychalcone induces autophagy through activation of the transcription factors TFE3 and TFEB.
3,4-Dimethoxychalcone induces autophagy through activation of the transcription factors TFE3 and TFEB.
EMBO molecular medicine (2019-10-15)
Guo Chen, Wei Xie, Jihoon Nah, Allan Sauvat, Peng Liu, Federico Pietrocola, Valentina Sica, Didac Carmona-Gutierrez, Andreas Zimmermann, Tobias Pendl, Jelena Tadic, Martina Bergmann, Sebastian J Hofer, Lana Domuz, Sylvie Lachkar, Maria Markaki, Nektarios Tavernarakis, Junichi Sadoshima, Frank Madeo, Oliver Kepp, Guido Kroemer
ABSTRACT
Caloric restriction mimetics (CRMs) are natural or synthetic compounds that mimic the health-promoting and longevity-extending effects of caloric restriction. CRMs provoke the deacetylation of cellular proteins coupled to an increase in autophagic flux in the absence of toxicity. Here, we report the identification of a novel candidate CRM, namely 3,4-dimethoxychalcone (3,4-DC), among a library of polyphenols. When added to several different human cell lines, 3,4-DC induced the deacetylation of cytoplasmic proteins and stimulated autophagic flux. At difference with other well-characterized CRMs, 3,4-DC, however, required transcription factor EB (TFEB)- and E3 (TFE3)-dependent gene transcription and mRNA translation to trigger autophagy. 3,4-DC stimulated the translocation of TFEB and TFE3 into nuclei both in vitro and in vivo, in hepatocytes and cardiomyocytes. 3,4-DC induced autophagy in vitro and in mouse organs, mediated autophagy-dependent cardioprotective effects, and improved the efficacy of anticancer chemotherapy in vivo. Altogether, our results suggest that 3,4-DC is a novel CRM with a previously unrecognized mode of action.
Sigma-Aldrich
Monoclonal Anti-ATG5 antibody produced in mouse, clone ATG5-18, purified immunoglobulin, buffered aqueous solution
Roche
Anti-HA High Affinity, from rat IgG1