Merck
  • Methoxymethyl Threofuranosyl Thymidine (4'-MOM-TNA-T) at the T7 Position of the Thrombin-Binding Aptamer Boosts Anticoagulation Activity, Thermal Stability, and Nuclease Resistance.
Methoxymethyl Threofuranosyl Thymidine (4'-MOM-TNA-T) at the T7 Position of the Thrombin-Binding Aptamer Boosts Anticoagulation Activity, Thermal Stability, and Nuclease Resistance.
ACS omega (2020-01-21)
Manojkumar Varada, Manisha Aher, Namrata Erande, Vaijayanti A Kumar, Moneesha Fernandes
ABSTRACT
The synthesis of 4'-methoxymethyl threofuranosyl (4'-MOM-TNA) thymidine and derived oligomers of the G-rich thrombin-binding aptameric (TBA) sequence is reported. The G-quadruplex stability, anticoagulation activity, and the enzymatic stability of these oligomers bearing the 2'-3'-phosphodiester backbone as single substitutions in the loop regions are studied. Amongst all the oligomers, TBA-7T bearing the 4'-MOM-TNA unit at the T7 position formed a quadruplex with the highest thermal stability. It also resulted in enhanced anticlotting activity that allowed a one-third reduction in the dose, relative to TBA. Further, TBA-7T exhibited enhanced nuclease resistance properties to both endo- and exonucleases.
Sigma-Aldrich
Fibrinogen from human plasma, 50-70% protein (≥80% of protein is clottable)