Merck
  • A large-scale viability assessment of the potato cryobank at the International Potato Center (CIP).
A large-scale viability assessment of the potato cryobank at the International Potato Center (CIP).
In vitro cellular & developmental biology. Plant : journal of the Tissue Culture Association (2017-11-07)
Rainer Vollmer, Rosalva Villagaray, José Cárdenas, Mario Castro, Oswaldo Chávez, Noelle L Anglin, David Ellis
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Molecular Biology, ≥70 Kunitz units/mg protein, lyophilized