Merck
  • Comparison of the thermal stability of lithiated graphite in LiBOB EC/DEC and in LiPF6 EC/DEC
Comparison of the thermal stability of lithiated graphite in LiBOB EC/DEC and in LiPF6 EC/DEC
Electrochemical and Solid-State Letters (2003)
Jiang J and Dahn JR