- A layer-built rechargeable lithium ribbon-type battery for high energy density textile battery applications
A layer-built rechargeable lithium ribbon-type battery for high energy density textile battery applications
Journal of Material Chemistry A (2014)
Kim J, et al.