Merck
  • Hydrogen and methane selectivity during alkaline supercritical water gasification of biomass with ruthenium-alumina catalyst
Hydrogen and methane selectivity during alkaline supercritical water gasification of biomass with ruthenium-alumina catalyst
Applied Catalysis. B, Environmental (2013)
Onwudili JA and Williams PT