Merck
  • Hydrogen generation using sodium borohydride solution and metal catalyst coated on metal oxide
Hydrogen generation using sodium borohydride solution and metal catalyst coated on metal oxide
International Journal of Hydrogen Energy (2002)
Kojima Y, et al.