- A comparison of sodium borohydride as a fuel for proton exchange membrane fuel cells and for direct borohydride fuel cells
A comparison of sodium borohydride as a fuel for proton exchange membrane fuel cells and for direct borohydride fuel cells
Journal of Power Sources (2006)
Wee JH