- A 3D heterometallic Pb(II)-Na(I) coordination polymer as a sensitive and selective fluorescent sensor for detecting Fe3 +
A 3D heterometallic Pb(II)-Na(I) coordination polymer as a sensitive and selective fluorescent sensor for detecting Fe3 +
Inorganic Chemistry Communications (2017)
Ming M and Bai N