Merck
  • Solid-state self-assembly of a complex from 1, 3, 5-benzenetricarboxylic acid and 1, 3, 5-trihydroxybenzene: influence of strong O-H--O and C-H--O hydrogen bonds
Solid-state self-assembly of a complex from 1, 3, 5-benzenetricarboxylic acid and 1, 3, 5-trihydroxybenzene: influence of strong O-H--O and C-H--O hydrogen bonds
New. J. Chem. (2001)
Liu R, et al.