Merck
  • Similar rates of chromosomal aberrant secondary oocytes in two indigenous cattle (Bos taurus) breeds as determined by dual-color FISH
Similar rates of chromosomal aberrant secondary oocytes in two indigenous cattle (Bos taurus) breeds as determined by dual-color FISH
Theriogenology (2012)
Pauciullo, Alfredo, et al