Merck
  • Osteogenic and odontogenic differentiation potential of dental pulp stem cells isolated from inflamed dental pulp tissues (I-DPSCs) by two different methods.
Osteogenic and odontogenic differentiation potential of dental pulp stem cells isolated from inflamed dental pulp tissues (I-DPSCs) by two different methods.
Acta odontologica Scandinavica (2019-12-20)
Vellore Kannan Gopinath, S Soumya, Manju Nidagodu Jayakumar
ABSTRACT
Objective: The objective of the present study is to isolate stem cells from inflamed dental pulp tissues (I-DPSCs) and study the characteristic such as surface markers, osteo/odontogenic differentiation potential between the outgrowth (OG) and enzymatic digestion (COL) methods.Materials and methods: I-DPSCs harvested by both methods were analysed for Mesenchymal Stem Cell marker expression by flow cytometry. The metabolic activity of the isolated cells was assessed by MTT assay. The Alkaline Phosphatase (ALP) and Alizarin red staining was done to analyse the osteogenic potential of isolated cells. The osteo/odontogenic differentiation was done by checking the expression of Dentine Matrix Protein 1 (DMP1), Dentine Sialophosphoprotein (DSPP), ALP and Bone Gamma-Carboxyglutamate Protein (BGLAP) by Real time PCR.Results: The isolated cells were positive for MSC markers such as CD-90, CD-105 and CD-73 and negative for CD-14, CD-45 and STRO-1. MTT assay indicated that the I-DPSCs from OG method showed higher metabolic activity than cells from COL. However, the osteo/odontogenic differentiation was in favour of cells isolated by COL method.Conclusion: Although the cell metabolic rate was more in OG, the osteo/odontogenic differentiation was higher in COL, suggesting that the isolation method and culture conditions do affect the differentiation capacity of isolated cells.
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)